Adapter, 75 Degree Side Arm, Ace Glass Incorporated
Supplier: Ace Glass
5045-10
89051-246EA 240.97 CAD
89051-246
Adapter, 75 Degree Side Arm, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a side arm at a 75° angle.
  • Top and side reinforced [ST] outer joints
  • Bottom [ST] inner joint
  • With side arm
  • Fabricated from borosilicate glass
This adapter comes with reinforced standard taper outer joints at the top and side, and an standard taper inner joint at the bottom. Side arm is at 75 degree angle.