NMR Tube Washer, Ace Glass Incorporated
Supplier: Ace Glass
2540-12 2540-13 2540-15 2540-19 2540-23 2540-25 2540-29 2540-31 2540-42 2540-54
89050-950EA 213.79 CAD
89050-950 89050-952 89050-954 89050-956 89050-958 89050-960 89050-962 89050-964 89050-966 89050-968
NMR Tube Washer, Ace Glass Incorporated
Tubes NMR Tubes
Rugged NMR tube washer designed with either Ace-Threds for ease and convenience or Ace-Thred and standard taper joints
The all-threaded version has a filter flask with a #15 Ace-Thred at the top for securing tube washer with a bushing, O-Ring, and #11 Ace-Thred side port with a removable 'Ace-Safe' hose connection for connecting to suction.
The threaded/jointed version has a filter flask with a standard taper 24/40 joint at top for connecting to inner standard taper joint at bottom of tube washer. It also features a serrated hose connection on the side for connecting to suction.