Nalgene® Large Centrifuge Bio Bottles with Sealing Cap, Polypropylene Copolymer, Thermo Scientific
Supplier: Thermo Fisher Scientific
3120-2006 3120-2024
89050-122CS 1106.32 CAD
89050-122 CA89094-358
Nalgene® Large Centrifuge Bio Bottles with Sealing Cap, Polypropylene Copolymer, Thermo Scientific
Bottles Centrifuge Bottles
Bottles are designed for use with the Sorvall® RC12BP centrifuge and H-12000 swing-bucket rotor.
Note: Before autoclaving, place the cap on the bottle without engaging the threads. For proper performance, fill the bottle to at least 80% of the total capacity.
They are designed to process large harvests and large volume gross separations in bacterial, yeast, and tissue isolations. These graduated bottles have smooth inner walls, excellent chemical resistance, and a 50-cycle life span. Polypropylene sealing caps are autoclavable. Bottles meet USP VI requirements. They are non-cytotoxicMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature range 4 to 22°C (39 to 72°F)