pH Indicator Strips, pH-Fix, Macherey-Nagel
Supplier: Macherey-Nagel
92110 92130.3 92115 92120.3
89049-924PK 52.77 CAD
89049-924 89049-926 89049-928 89049-930
pH Indicator Strips, pH-Fix, Macherey-Nagel
Test Papers pH Indicator Papers and Test Strips
pH-Fix test strips allow fast pH testing directly at the point of interest.
  • Ready to use
  • Easy and safe
  • Packed in classic flat box
  • Several test pads for exact results
  • Safe analysis by long plastic handle
  • Fixed indicator dye does not leach into sample
Even in weakly buffered solutions, a safe and reliable pH determination becomes possible, as the strips can be immersed for a long time until the final reaction color has developed
pH-Fix test strips are tested and approved for medical applications and carry a CE-mark for in-vitro-diagnostics 98/79/EGMore Product Information