AC Medium Rounds, Clear, Wide Mouth, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
5710133V-21 5710238V-21 5710448V-21 5710858V-21 5711670V-21 5710133C-21 5710238C-21 5710448C-21 5710858C-21 5711670C-21 5710238V-22 5710448V-22 5710133C-22 5710238C-22 5710448C-22 5710858C-22 5711670C-22 5710238V-25 5710133C-25 5710238C-25 5710133C-26 5710238C-26 5710448C-26 5710858C-26 5711670C-26 5710133B 5710238B 5710448B 5710858B 5711670B
89046-344CS 155.86 CAD
89046-344 89046-346 89046-348 89046-350 89046-352 89046-354 89046-356 89046-358 89046-360 89046-362 89046-366 89046-368 89046-374 89046-376 89046-378 89046-380 89046-382 89046-426 89046-428 89046-430 89046-442 89046-444 89046-446 89046-448 89046-450 89046-452 89046-454 89046-456 89046-458 89046-460
AC Medium Rounds, Clear, Wide Mouth, Kimble®, DWK Life Sciences
Bottles Glass Bottles
Versatile storage bottles provide an ideal containment for solid, liquid, or powdered specimens and samples.
The clear Type III soda-lime glass construction allows for constant sample viewing. Wide mouth design ensures efficient sample addition and removal. Bottles are supplied with screw-on caps, unless otherwise noted.
Ordering information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 to 48 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 432 per wrap) packaged in corrugated cases. Bottle caps areMore Product Information