VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
89079-490 89079-494 89039-656 89039-658 89039-660 89039-662 89039-664 89039-666 89039-668 89039-670 89079-492 89401-562 89401-564 89401-566 89401-568
89079-490CS 337.22 CAD
89079-490 89079-494 89039-656 89039-658 89039-660 89039-662 89039-664 89039-666 89039-668 89039-670 89079-492 89401-562 89401-564 89401-566 89401-568
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Tubes Centrifuge Tubes
These disposable conical-bottom and freestanding tubes feature black printed graduations and smooth inner walls for easy filling and sample preparation.
  • Easy-to-read black graduations in ±2% increments
  • White printed writing area for permanent coding
  • RNase-, DNase-, and endotoxin-free
  • Autoclavable (121 °C) and freezable (–80 °C)
  • Can be centrifuged up to 12500×g
  • Sterile tubes are sterilized by irradiation
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters