VWR® Natural Gas Micro Burner
Supplier: VWR International
89038-526
89038-526CS 1946.95 CAD
89038-526
VWR® Natural Gas Micro Burner
Heaters Gas Burners
Micro burner with flame stabilizer provides an intense small flame suitable for micro procedures and analyses.
  • BTU output: 800–1200
  • Fuel type: Natural gas
Sturdy die-cast base is constructed from nickel-plated zinc-alloy with 8mm (5/16") serrated gas inlet, 13mm (1/2") aluminum mixing tube, and adjustable orifice. Needle valve simplifies air/gas adjustment.