Unifrost™ Microscope Slides, Azer Scientific
Supplier: Azer Scientific Inc.
EMS200U
89033-048CS 790.48 CAD
89033-048
Unifrost™ Microscope Slides, Azer Scientific
Slides Microscope Slides
Manufactured from high-quality white German glass, with a single frosted end.
  • Single frosted end to help prevent sticking
  • Colored end helps with quick identification and organization
All slides have single side, color frosted ends which helps to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory.
Order Now
SPECIFICATIONS
Environmentally Preferable