Amplitude™ Theta™ Wipes, Contec®
Supplier: Contec
AMTH0001 AMTH0002
89030-190CS 532.77 CAD
89030-190 89030-192
Amplitude™ Theta™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Theta™ Wipes are made from a lightweight hydroentangled cellulose/polyester, which creates an economical absorbent wipe ideal for general cleaning applications.
  • Recommended for ISO Class 6 to 8 environments
  • Economical and cost-effective nonwoven wipe
  • Lightweight sorbent wipe
  • Double packaged to preserve product integrity
These wipes offer excellent durability and sorbency. They are designed for nonabrasive applications, such as general-purpose wiping and polishing of surfaces.
Order Now
SPECIFICATIONS
Environmentally Preferable