Amplitude™ EcoCloth™ Wipes, Contec®
Supplier: Contec
AMEC0001 AMEC0002 AMEC0003 AMEC0004 AMEC0005
89030-164CS 922.08 CAD
89030-164 89030-166 89030-168 89030-170 89030-172
Amplitude™ EcoCloth™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The EcoCloth™ wipe has been engineered to provide greater thickness in a lighter weight material. Amplitude™ EcoCloth™ is a hydroentangled cellulose/polyester fabric with additional loft. It is lightweight but has excellent sorbency which exceeds comparable standard cellulose/polyester wipes.
  • Recommended for ISO Class 5/6 environments
  • Low basis weight cellulose/polyester wipe with additional loft for greater sorbency
  • Most sorbent of all cellulose/polyester wipes
  • Lighter weight with the same performance, wipes can reduce solid waste in excess of 40%
  • Double
More Product Information