Flow Control Adapters, Straight, with PTFE Stopcock, Chemglass
Supplier: Chemglass
CG-1029-01 CG-1029-02 CG-1029-29 CG-1029-14 CG-1029-19
80065-884EA 243.61 CAD
80065-884 80065-886 80065-892 89028-202 89028-204
Flow Control Adapters, Straight, with PTFE Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow control adapters, used to regulate the flow of gases or liquids into reaction systems.
  • Equipped with PTFE stopcock
PTFE stopcock has a bore of 2 mm. Hose connection has an O.D. of 8 mm at the middle serration.
Order Now
SPECIFICATIONS
Environmentally Preferable