C Mag HS 7 Hot Plate Stirrer, IKA® Works
Supplier: IKA Works
0003581201
89028-160EA 2832.09 CAD
89028-160
C Mag HS 7 Hot Plate Stirrer, IKA® Works
Stirrers Magnetic Hotplate Stirrers
Hot plate stirrer features a digital temperature display, microprocessor control, and extremely fast heating. Fixed safety circuit at 550 °C (1022 °F).
Ceramic top plate measures 17.8x17.8cm (7x7"). May be used with temperature sensor 89028-114.