Sklar Edge™ Tungsten Carbide Mayo Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
16-1505 16-1515 16-1605 16-1615 16-1520 16-1525 16-1620 16-1625 16-3530 16-3531 16-3532 16-3533
89025-668EA 619.01 CAD
89025-668 89025-672 89025-674 89025-678 76192-038 76192-040 76192-042 76192-044 76192-066 76192-068 76192-070 76192-072
Sklar Edge™ Tungsten Carbide Mayo Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Mayo Dissecting Scissors are a modification of standard Mayo scissors that provide an exceptionally hard and durable cutting edge designed to stay sharp for a significantly longer period of time than standard scissors
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar permanently bonds the tungsten carbide inserts to the blade of the scissors whichMore Product Information