Econo™ Sterile Lorna (Edna) Towel Clamps, Sklar
Supplier: Sklar
96-2528 96-2529
89023-806CS 1196.78 CAD
89023-806 89023-808
Econo™ Sterile Lorna (Edna) Towel Clamps, Sklar
Clamps General Purpose Clamps
Econo™ Sterile Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site.
  • This clamp should never be used on skin
  • Ready-to-use, sterile, disposable, floor-grade instruments
The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firmMore Product Information