Surgi-OR™ Lister Bandage Scissors, Physician Grade, Sklar
Supplier: Sklar
95-222 95-231 95-241
89023-524EA 52.26 CAD
89023-524 89023-528 89023-532
Surgi-OR™ Lister Bandage Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ Lister Bandage Scissors are a versatile instrument most commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord.
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterusMore Product Information