Toteline Nest and Stack Container, MFG Tray
Supplier: Molded Fiber Glass Tray
780008W-5269 780108W-5269 780118-5269 780208-5269 780218-5269 780308-5269 780318-5269 780408-5269 780418-5269 780508-5269 780608-5269 780808-5269
89022-842EA 384.02 CAD
89022-842 89022-844 89022-846 89022-850 89022-852 89022-856 89022-858 89022-862 89022-864 89022-868 89022-874 89022-886
Toteline Nest and Stack Container, MFG Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
MFG Tray's durable Nest and Stack containers are resist to cutting oils, grease, and solvents
Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for work-in-process part handling. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery,More Product Information