Diamond-Plate Electrically Conductive Runner, Wearwell®
Supplier: Wearwell
785.532X3X75DPBK
89021-406RL 7886.65 CAD
89021-406
Diamond-Plate Electrically Conductive Runner, Wearwell®
Floor Mats Permanent Floor Mats
Electrically Conductive Runner is the ideal runner for areas where static prevention is a priority.
  • Treated surface drains static electricity from personnel
  • Protect sensitive equipment from damage or eliminate static annoyance
  • Grounding snap is attached to the corner of each mat
The Electrically Conductive Runner can be rolled out for long aisle ways or cut to size for specific workstations. Use the Electrically Conductive Runner to protect sensitive equipment or eliminate static annoyance. A grounding snap is included on each mat, and grounding isMore Product Information