Diamond-Plate SpongeCote™ and UltraSoft Floor Mats, Wearwell®
Supplier: Wearwell
415.916X3X75BK 415.916X3X75BYL 415.916X2X75BK 415.916X2X75BYL 415.916X2X75CHV 415.916X2X3BK 415.916X2X75GY 415.916X2X3GY 415.916X2X3BYL 415.916X3X75CHV 415.916X3X75GY 415.916X3X5GY 415.916X3X5BYL 415.916X4X75BK 415.916X4X75CHV 415.916X4X75GY 415.916X4X75BYL 415.916X2X3CHV 415.916X3X5CHV 414.1516X2X3GY 414.1516X2X3BYL 414.1516X2X75BK 414.1516X2X75GY 414.1516X2X75BYL 414.1516X3X5BK 414.1516X3X5GY 414.1516X3X5BYL 414.1516X3X75BK 414.1516X3X75GY 414.1516X3X75BYL 414.1516X4X75BK 414.1516X4X75GY 414.1516X4X75BYL 415.916X6X75BYL 415.916X6X75BK 415.916X5X75BYL 415.916X5X75BK 415.916X3X5BK
89020-386RL 4424.02 CAD
89020-386 89020-388 89020-390 89020-392 89020-394 89020-396 89020-398 89020-400 89020-402 89020-404 89020-408 89020-410 89020-412 89020-414 89020-416 89020-418 89020-420 89020-422 89020-424 89020-778 89020-782 89020-784 89020-786 89020-790 89020-792 89020-794 89020-798 89020-800 89020-802 89020-806 89020-808 89020-810 89020-814 89020-816 89021-448 89021-450 89021-452 89021-454 89020-406
Diamond-Plate SpongeCote™ and UltraSoft Floor Mats, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
These industrial strength, anti-fatigue diamond-plate mats are designed for comfort and multiple shift operation use
  • Industrial strength
  • Antifatigue
  • Resists abrasion and most chemical spills
The SpongeCote™ mats feature a 1cm (3/8") thick Nitricell® sponge base and have an overall thickness of 1.4cm (9/16"). The UltraSoft mats feature a double-thick Nitricell® sponge base and have an overall thickness of 2.4cm (15/16").
The diamond-plate surface is designed for heavy-use production and lab areas, and resists abrasion and most chemical spills. TheMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable