SoftStep Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
427.38X2X60BK 427.38X2X60BYL 427.38X2X3BK 427.38X2X3BYL 427.38X3X60BK 427.38X3X60BYL 427.38X3X5BYL 427.38X3X12BYL 427.38X3X5BK 427.38X3X12BK 427.38X4X60BK 427.38X4X60BYL
89020-554RL 1158.25 CAD
89020-554 89020-558 89020-560 89020-564 89020-566 89020-568 89020-572 89020-574 89020-576 89020-578 89020-584 89020-588
SoftStep Anti-Fatigue Mat, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
When the budget is tight, but leg and back fatigue must be battled, try SoftStep.
  • Economic comfort for light industrial applications
  • Safety rounded edges
  • Ribbed, pebble, and diamond surface patterns
  • Standard lengths or easily cut to fit on-site
SoftStep provides a no frills PVC blown sponge barrier from hard floors. With an attractive pebbled surface, this mat can be purchased in rolls and easily cut to length on-site for quick replacement when damaged. Ideally suited for use in dry environments.
Certifications: Meets ASTM F1677. Dry: COF = 0.60.
Order Now
SPECIFICATIONS
Environmentally Preferable