Automatic Digital Refractometers, ATAGO®
Supplier: Atago
3261RX5000A 3921 3122 3263 3271
47752-846EA 44446.9 CAD
47752-846 89008-790 89008-808 89008-810 97015-530
Automatic Digital Refractometers, ATAGO®
Refractometers Benchtop Refractometers
Digital refractometers offer automatic measurement of the refractive index and Brix scale.
Model RX-007[alpha] is designed for measuring water soluble samples with very low concentrations (5% or less), such as tea and low sugar beverages. It has a measurement temperature of 15 to 30°C (59 to 86°F). Models RX-5000[alpha], RX-7000[alpha], and RX-9000[alpha] are equipped with a Peltier thermo module for maintaining a constant measuring temperature. Measurement starts after the sample reaches the target temperature. Up to 30 user scales can be programmed on these models, and they also featureMore Product Information