polyproLABS® Wall Cabinets, Air Control
Supplier: Air Control
AC-240-3030 AC-240-3630 AC-240-4230 AC-240-4830 AC-240-3024 AC-240-3624 AC-240-4224 AC-240-4824 AC-250-3030 AC-250-3048 AC-250-3630 AC-250-4230 AC-250-4830 AC-250-3024 AC-250-3624 AC-250-4224 AC-250-4824 AC-205-3030 AC-205-3630 AC-205-4230 AC-205-4830 AC-205-3024 AC-205-3624 AC-205-4224 AC-205-4824 AC-230-1830 AC-230-2430 AC-230-3030 AC-230-3630 AC-230-1824 AC-230-2424 AC-230-3024 AC-230-3624 AC-200-1830 AC-200-2430 AC-200-3030 AC-200-3630 AC-200-1824 AC-200-2424 AC-200-3024 AC-200-3624 AC-210-3030 AC-210-3630 AC-210-4230 AC-210-4830 AC-210-3024 AC-210-3624 AC-210-4224 AC-210-4824 AC-210-4848 AC-220-3030 AC-220-3630 AC-220-4230 AC-220-4830 AC-220-3036 AC-220-3636 AC-220-4236 AC-220-4836 AC-220-5430 AC-220-7236 AC-215-3030 AC-215-5030 AC-215-4230 AC-215-4830 AC-215-3024 AC-215-3624 AC-215-4224 AC-215-4824
89008-514EA 4396.1 CAD
89008-514 89008-516 89008-518 89008-520 89008-522 89008-524 89008-526 89008-528 89008-530 89008-532 89008-534 89008-536 89008-538 89008-540 89008-542 89008-544 89008-546 89008-548 89008-550 89008-552 89008-554 89008-556 89008-558 89008-560 89008-562 89008-564 89008-566 89008-568 89008-570 89008-572 89008-574 89008-576 89008-578 89008-580 89008-582 89008-584 89008-586 89008-588 89008-590 89008-592 89008-594 89008-596 89008-598 89008-600 89008-602 89008-604 89008-606 89008-608 89008-610 89008-612 89008-614 89008-616 89008-618 89008-620 89008-622 89008-624 89008-626 89008-628 89008-630 89008-632 89008-746 89008-748 89008-750 89008-752 89008-754 89008-756 89008-758 89008-760
polyproLABS® Wall Cabinets, Air Control
Furniture Casework
Corrosion-resistant casework cabinets are constructed of stress-relieved, acid-resistant, 1.3 cm (1/2") thick polypropylene
Frameless acrylic sliding doors are 1 cm (3/8") thick. Framed acrylic doors have polypropylene frames and 1 cm (3/8") thick acrylic panels with beveled edges. Solid polypropylene doors are 1 cm (3/8") thick with beveled edges.
Cabinets include an adjustable shelf, which is made from solid 1.3 cm (1/2") thick polypropylene. Blind corner cabinets do not include a shelf. Open-front bookcase cabinets include two adjustable shelves.
Joints are thermally welded by aMore Product Information