Hydrovoid Desiccator Cabinets, Air Control
Supplier: Air Control
4311-0 4321-1 4321-2 4331-1 4331-2 4341-1 4341-2 4351-1 4351-2 4361-1 4361-2 4381-1 4381-2 43101-1 4411-0 4421-1 4421-2 4431-1 4431-2 4441-1 4441-2 4451-1 4451-2 4461-1 4461-2 4481-1 4481-2 44101-1 4511-0 4521-1 4521-2 4531-1 4531-2 4541-1 4541-2 4551-1 4551-2 4561-1 4561-2 4581-1 4581-2 45101-1 4611-0 4621-1 4621-2 4631-1 4631-2 4641-1 4641-2 4651-1 4651-2 4661-1 4661-2 4681-1 4681-2 46101-1
89008-264EA 2028.61 CAD
89008-264 89008-266 89008-268 89008-270 89008-272 89008-274 89008-276 89008-278 89008-280 89008-282 89008-284 89008-286 89008-288 89008-290 89008-292 89008-294 89008-296 89008-298 89008-300 89008-302 89008-304 89008-306 89008-308 89008-310 89008-312 89008-314 89008-316 89008-318 89008-320 89008-322 89008-324 89008-326 89008-328 89008-330 89008-332 89008-334 89008-336 89008-338 89008-340 89008-342 89008-344 89008-346 89008-348 89008-350 89008-352 89008-354 89008-356 89008-358 89008-360 89008-362 89008-364 89008-366 89008-368 89008-370 89008-372 89008-374
Hydrovoid Desiccator Cabinets, Air Control
Desiccators Desiccator Cabinets
Cabinets are designed for protection and storage of biological mixtures, particulate-sensitive semiconductor parts, micro-electronic devices, and electronic sub-assemblies
Cabinets are constructed to allow clear visibility of contents without opening doors. Tight, solvent-fused seams permit easy cleaning and heavy, non-rusting hardware ensures a long life. Closed-cell neoprene gasketing around door assemblies and multiple 90° locking knobs provide dust-free, gas-tight storage. Cabinets have one door per chamber. Standard units are available in clear or UV-shielding amber acrylic. ESD-safeMore Product Information