KIMAX® Cold Test Jar, Cloud and Pour Points, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
32501-99
89002-138CS 1210.6 CAD
89002-138
KIMAX® Cold Test Jar, Cloud and Pour Points, Kimble Chase
Petroleum Testing Apparatus
Used to determine the cloud point of petroleum oils and the point at which chilled, undisturbed oils will pour
Flat bottom jar features a reinforced bead at open end, a marking spot, and a graduation ring at 54mm (21/8") from inside bottom.
Certifications: Meets ASTM D2500 and D97 requirements.