KIMAX® Gas Sampling Tube, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
30040-500
89002-136CS 1230.94 CAD
89002-136
KIMAX® Gas Sampling Tube, Kimble Chase
Tubes Gas Sampling Tubes
Gas collecting apparatus features tubulations at each end that accept 9.5 mm (³/₈") flexible tubing.