KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Class B, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28015P-100
89002-006CS 953.56 CAD
89002-006
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Class B, Kimble Chase
Flasks Volumetric Flasks
Flasks have a graduation ring blasted on the neck
Designed from ASTM specification E288, Class B requirements.
Calibrated to contain. Provided with yellow and blue closed-bottom linear, high-density polyethylene stopper to fit the [ST] neck. Enlarged stopper top protects the flask neck if tipped over.