KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28014F-10 28014F-2 28014F-5 28014F-100 28014F-1000 28014F-200 28014F-2000 28014F-25 28014F-250 28014F-50 28014F-500
89001-940CS 1072.78 CAD
89001-940 89001-946 89001-956 89001-942 89001-944 89001-948 89001-950 89001-952 89001-954 89001-958 89001-960
KIMAX® Volumetric Flasks with Color-Coded [ST] PTFE Stopper, Class A, Kimble Chase
Flasks Volumetric Flasks
These flasks are calibrated to contain and feature a graduation ring blasted on the neck
Sizes 10mL and larger feature a marking area. Supplied with a [ST] PTFE stopper with color-coded handle. The 2mL size is shaped like a test tube; all other sizes are of a conventional flask shape. Sizes 5mL and larger are designed from ASTM Specification E288, Class A unserialized requirements.