KIMAX® Single Metric Scale Graduated Cylinders, Class B, Low Form, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20032-50 20032-100
89001-200CS 1491.33 CAD
89001-200 89001-202
KIMAX® Single Metric Scale Graduated Cylinders, Class B, Low Form, Kimble Chase
Cylinders Measuring Cylinders
Calibrated "to deliver." Cylinders are made of sturdy borosilicate glass and feature two pour spouts
Bodies have a larger diameter with lower overall height than comparable standard cylinders, providing greater stability and decreased breakage. Scale is durable white ceramic enamel.
Order Now
SPECIFICATIONS
Form Short
Environmentally Preferable