KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
14020-400 14020-1000 14020-100 14020-300 14020-600 14030-100 14030-600 14030-1000 14030-200 14030-300 14030-400
13920-034PK 178.18 CAD
13920-034 13920-078 13920-137 13920-012 13920-057 89000-698 89000-748 89000-750 89000-752 89001-086 89001-088 89001-090
KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Beakers
These berzelius, tall form, double capacity scale beakers are manufactured with uniform wall thickness, particularly in the critical radius point of the heel area.
  • Beakers feature a slightly flared top with a thick reinforcing bead
  • Available with or without a pouring spout
  • All sizes have a durable matte finish area for clear marking, and easy-to-read white scales to indicate approximate volumes
This strong and durable beaker exhibits excellent mechanical and thermal shock resistance.
Certifications: Beakers with a pouring spout are designed fromMore Product Information