KIMAX® Graduated Cylinders, Class B, To Contain, with Glass Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
20039-10 20039-500 20039-2000 20039-25 20039-50 20039-100 20039-250 20039-1000
89000-760CS 2044.48 CAD
89000-760 89000-762 89000-764 89001-204 89001-206 89001-208 89001-210 89001-212
KIMAX® Graduated Cylinders, Class B, To Contain, with Glass Stoppers, Kimble Chase
Cylinders Measuring Cylinders
Cylinders and [ST] stoppers are made of sturdy borosilicate glass
The 250mL cylinder may be used to determine unsaponified matter in soaps and soap products (ASTM D460), and anhydrous salt free soda soap and fatty matter in soaps containing synthetic detergents (ASTM D820). The 500mL cylinder may be used in settlement tests of emulsified asphalts (ASTM D244).
Cylinders are designed from ASTM Specification E1272, Style 2, Class B requirements. Stoppers are designed from ASTM Specification E675.
Single metric scale is in durable white ceramic enamel. The 10mL and 25mL cylinders areMore Product Information
Order Now
SPECIFICATIONS
Form Short
Environmentally Preferable