Purifier® Vertical Clean Benches, Labconco®
Supplier: Labconco
3970203 3970204 3970223 3970224 3970303 3970304 3970323 3970324 3970400 3970401 3970403 3970404 3970420 3970421 3970423 3970424 3970200 3970201 3970220 3970221 3970300 3970301 3970320 3970321
89000-048EA 14362.2 CAD
89000-048 89000-050 89000-052 89000-054 89000-056 89000-058 89000-060 89000-062 89000-064 89000-066 89000-068 89000-070 89000-072 89000-074 89000-076 89000-078 89000-130 89000-132 89000-134 89000-136 89000-138 89000-140 89000-142 89000-144
Purifier® Vertical Clean Benches, Labconco®
Fume Hoods and Enclosures Laminar Flow Cabinets
Clean benches provide product protection from environmental contaminants for applications requiring a particulate-free work area, including plant tissue culture, electronic part inspection, syringe filling, medical device assembly, media plate preparation, and PCR
Base stands (sold separately) are made of durable 4.4 cm (1³/₄") thick, tubular, glacier white, epoxy-coated steel. The height of base stands is fully adjustable from 69.9 to 85.1 cm (27¹/₂ to 33¹/₂") in 2.5 cm (1") increments. Base stands with fixed feet come with leveling feet for use on uneven floor surfaces. Base stands withMore Product Information