VWR® PureStep Adhesive Floor Mats
Supplier: VWR International
82028-392 82028-394 82029-678 82029-680 82031-466 82031-676 87004-000 87004-002 87004-004 87004-006 87004-008 87004-012 87004-014 87004-016 87004-018 87004-020 87004-022 87004-024 87004-026 87004-028 87004-030 87004-032 87004-034 87004-036 87004-038 87004-040 87004-042 87004-046 87004-048 87004-050 87004-054 87004-056 87004-058 87004-060 87004-062 87004-064 87004-066 87004-068 87004-070 87004-072 87004-074 87004-076 87004-078 87004-080 87004-082 87004-084 87004-086 87004-088 87004-090 87004-092 87004-296 87004-298 87004-300 89066-232 89089-166 89126-760 89126-762 89126-766 89126-768 89126-770 89126-772 89131-500 87004-044 87004-052 89126-764 89428-992
82028-392CS 736.13 CAD
82028-392 82028-394 82029-678 82029-680 82031-466 82031-676 87004-000 87004-002 87004-004 87004-006 87004-008 87004-012 87004-014 87004-016 87004-018 87004-020 87004-022 87004-024 87004-026 87004-028 87004-030 87004-032 87004-034 87004-036 87004-038 87004-040 87004-042 87004-046 87004-048 87004-050 87004-054 87004-056 87004-058 87004-060 87004-062 87004-064 87004-066 87004-068 87004-070 87004-072 87004-074 87004-076 87004-078 87004-080 87004-082 87004-084 87004-086 87004-088 87004-090 87004-092 87004-296 87004-298 87004-300 89066-232 89089-166 89126-760 89126-762 89126-766 89126-768 89126-770 89126-772 89131-500 87004-044 87004-052 89126-764 89428-992
VWR® PureStep Adhesive Floor Mats
Floor Mats Tacky Mats
Multilayered contamination control mats remove and contain dirt and dust from foot traffic and equipment wheels.
  • 0.3 mil thick acrylic-based, pressure-sensitive adhesive coating
  • Antimicrobial agent protects against growth of bacteria, mold, and fungus
Mats consist of 30 or 60 sheets of 1.5 mil low-density polyethylene film. Each sheet has a 0.3 mil thick acrylic-based, pressure-sensitive adhesive coating. Numbered tabs ensure one-at-a-time sheet removal. Mats can easily be removed during routine floor maintenance. Some sizes are available with high-More Product Information