VWR® Signature™ Single Channel Pipettors, Fixed Volume
Supplier: VWR International
83009-758 83009-760 83009-762 83009-764 83009-766 83009-768 83009-770 83009-774 83009-776 83009-772
83009-758EA 853.69 CAD
83009-758 83009-760 83009-762 83009-764 83009-766 83009-768 83009-770 83009-774 83009-776 83009-772
VWR® Signature™ Single Channel Pipettors, Fixed Volume
Pipettes Micropipettes
Ideal for applications where specific volumes are used repeatedly, these slender, contoured pipettor bodies fit comfortably in either hand, as well as in large or small hands.
  • Ten fixed volumes available
  • Narrow shaft for universal tip fit
  • Removable, adjustable tip ejector
  • Fully autoclavable
  • VWR® two-year limited warranty
  • Pipettor stands hold six pipettors
In addition, maintenance-free seals and highly polished pistons provide exceptional accuracy and precision, as well as allow the pipettors to be autoclaved repeatedly without recalibration