VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Supplier: Brandtech
91789 91989 92089
83009-050PK 274.17 CAD
83009-050 83009-052 83009-053 83009-054
VITLAB® Square Laboratory Bottles, HDPE, Narrow Mouth, with PP Screw Caps, BrandTech
Bottles Plastic Bottles
General-use, high-density polyethylene laboratory bottles come with polypropylene screw caps.
  • Space-saving shape
  • HDPE construction with PP screw caps
  • Narrow mouth
Bottles provide excellent chemical resistance to aliphatic and aromatic hydrocarbons, alcohols, aldehydes, alkalis, and acids.
Order Now
SPECIFICATIONS
Environmentally Preferable