Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
604160 606160 760160 768160 604181 607180 710107 710160 710183 607160 760107 604107 606107 606180 607107 710180 760180 768180 604160-TRI 606160-TRI 607160-TRI 710160-TRI 760160-TRI 768160-TRI
82050-000CS 741.67 CAD
82050-000 82050-002 82050-072 82050-074 82050-474 82050-482 82051-116 82051-584 89085-300 82050-006 82050-070 82050-472 82050-476 82050-478 82050-480 82051-118 82051-182 82051-212 75799-848 75799-850 75799-852 75799-860 75799-862 75799-864
Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Pipettes Serological Pipettes
Pipettes are manufactured from optical-grade polystyrene and feature a high internal and external surface polish, which reduces meniscus formation and helps ensure complete liquid expulsion.
  • Polystyrene construction
  • Polish reduces meniscus formation
  • Free of RNase, DNase, and pyrogens
  • Designed to fit automatic, semiautomatic, and manual pipetting devices
Fine-line graduation printing further contributes to accuracy of at least ±2%. Each disposable pipette is cotton-plugged and color-coded to facilitate identification. Pipettes with ascending andMore Product Information