1536-Well µClear® Bottom Plates, Greiner Bio-One
Supplier: Greiner Bio-One
782095 782096 782097 783096
82051-402CS 2934.49 CAD
82051-402 82051-404 82051-406 82051-432
1536-Well µClear® Bottom Plates, Greiner Bio-One
Microplates HTS Microplates
Plates are manufactured from virgin polystyrene resin and feature µClear® microthin polystyrene film on the bottom.
  • Virgin polystyrene construction
  • Free of RNase, DNase, human DNA, and pyrogens
  • Designed to reduce air bubble formation in wells
  • Stackable with and without lids
Designed to be compatible with microplate handling equipment and robotics
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
The square-round well geometry is designed to minimize wicking and reduce air bubble formation, while increasing volume capacityMore Product Information