96-Well ELISA Strip Plates, Greiner Bio-One
Supplier: Greiner Bio-One
701001 701070 701071 701185 705070 705071 705073 705074 705075 705076 705185 754061 756001 756070 756071 762001 762061 762070 762071 762074 762075 762076 762077 767001 767070 767071 705091
82051-092CS 442.15 CAD
82051-092 82051-096 82051-098 82051-100 82051-102 82051-104 82051-106 82051-108 82051-110 82051-112 82051-114 82051-164 82051-166 82051-174 82051-178 82051-180 82051-184 82051-186 82051-188 82051-190 82051-192 82051-194 82051-196 82051-198 82051-204 82051-208 82051-210 82051-536 10027-748
96-Well ELISA Strip Plates, Greiner Bio-One
Microplates Immunology Microplates and Strips
Greiner Bio-One 96-well strip plates are designed for diagnostic and immunological research applications.
  • Strips mounted in frame
  • Virgin polystyrene construction
  • Chimney-well design helps reduce cross-contamination
  • Strip plates available in white, black, or clear formats
  • Free of RNase, DNase, and DNA-polymerase inhibitors
  • Nonpyrogenic
Microplates in strip format offer the advantage of greater flexibility in diagnostics
Quality controlled by means of ELISA. Clear MICROLON® products are tested in a colorimetric immunoassay, black FLUOTRAC™ plates inMore Product Information