CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Supplier: Greiner Bio-One
657165 657185 662102 662165 665102 665165 677102 677165 657160 662160 665180 677180
82050-844CS 217 CAD
82050-844 82050-846 82050-888 82050-894 82050-926 82050-928 82051-000 82051-002 82050-842 82050-892 82050-930 82051-004
CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Microplates Cell Culture Plates
Cell Culture Multiwell Plates allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Chimney well design helps reduce cross-contamination
Surface treatments provide the conditions for improved cell adherence, accelerated cell expansion, and they increase overall cell yield.
Manufactured from optically clear virgin polystyrene, these flat-bottom plates are supplied sterile and pyrogen-free with a printed lotMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable