CELLSTAR© Suspension Culture Flasks, Screw Cap, Hydrophobic Surface, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
658190 658195 660190 661190 661195 690190 690195
82050-858CS 270.16 CAD
82050-858 82050-860 82050-874 82050-880 82050-882 82051-076 82051-078
CELLSTAR© Suspension Culture Flasks, Screw Cap, Hydrophobic Surface, Sterile, Greiner Bio-One
Flasks Cell Culture Flasks
These graduated, canted-neck flasks are manufactured in an aseptic, fully automated facility from high-quality virgin polystyrene resin to ensure superior optical clarity.
  • High-quality virgin polystyrene construction
  • Superior optical clarity
  • Hydrophobic surface
  • Ideal for suspension cultures and hybridoma
Flask design makes it possible to efficiently reach the cell lawn with a cell scraper, while still providing optimal access with a pipet
All flasks are integrity tested to provide uniform growth and proliferation, and are sterilized by radiation.More Product Information