96-Well Microplates, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
650203 650204 650205 650206 650209 650261 651203 651205 651206 651209 651261 655204 655205 655206 655209 655261 650207 650201 651201 655201
82050-638CS 354.37 CAD
82050-638 82050-640 82050-642 82050-644 82050-646 82050-648 82050-668 82050-670 82050-672 82050-674 82050-676 82050-678 82050-778 82050-780 82050-782 82050-784 82050-786 89041-278 82050-636 82050-662 82050-774
96-Well Microplates, Polypropylene, Greiner Bio-One
Microplates HTS Microplates
These 96-well microplates are manufactured from ClearChoice™ virgin polypropylene resin and are produced with uniform external dimensions and tolerances to meet ANSI recommendations for compatibility with equipment and robotics.
  • Virgin polypropylene construction
  • Alphanumeric coding
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Stackable
  • Autoclavable
These plates are biologically inert, exhibit low binding characteristics, offer high temperature tolerance (–196 to 121 °C/–321 to 250 °F), and are resistant to many standard laboratory chemicals