96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
Supplier: Greiner Bio-One
655074 655075 655076 655077 655095 655096 655097 655094
82050-724CS 184.66 CAD
82050-724 82050-726 82050-728 82050-730 82050-752 82050-754 82050-756 82050-750
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
Microplates HTS Microplates
Black and white microplates are manufactured from virgin polystyrene resin and are designed to meet industry standard dimension specifications to facilitate compatibility with microplate handling and robotics.
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Virgin polystyrene construction
  • Solid bottoms or clear bottoms
All of the black and white plates have square chimney wells with flat bottoms
LUMITRAC™ and FLUOTRAC™ formats are offered to allow a wide variety of applications, including cell-based assays. White LUMITRAC™ plates are forMore Product Information