ThinCert™ Tissue Culture Inserts, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
657640 657641 657630 657631 657638 662640 662631 662638 665630 665638 657610 665610 662610 662641 662630 665640 665641 665631
82050-010CS 212.95 CAD
82050-010 82050-012 82050-014 82050-016 82050-018 82050-020 82050-026 82050-028 82050-034 82050-038 82051-568 82051-570 82051-572 82050-022 82050-024 82050-030 82050-032 82050-036
ThinCert™ Tissue Culture Inserts, Sterile, Greiner Bio-One
Microplates Insert Plates and Inserts
Tissue culture inserts provide optimal support for co-cultivation, cell migration, and transport process applications.
  • Sterile
  • Spacers eliminate capillary effect
  • Individually wrapped inserts
  • Low protein binding capacity
  • Improved cell adhesion
Advanced design permeable inserts feature transparent or translucent polyethylene terephthalate (PET) membranes that offer low protein binding capacity, improved cell adhesion, and defined pore size to ensure complete exchange of compounds and nutrients
ThinCert™ membranes are sealed to a robust, transparentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable