Backhaus Towel Clamp, Sklar
Supplier: Sklar
47-2935 47-2945 47-2955 47-2960
82030-262EA 101.33 CAD
82030-262 82030-264 82030-266 82030-268
Backhaus Towel Clamp, Sklar
Clamps General Purpose Clamps
Backhaus Towel Clamp is a perforating clamp used for grasping tissue, securing towels or drapes and holding or reducing small bone fractures.
Sklar Backhaus Towel Clamps are stainless steel, industry standard.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
SPECIFICATIONS
Environmentally Preferable
Disposable / reusable Reusable
Latex/Latex-free Latex-free