VWR® Freezer Racks
Supplier: VWR International
82024-518 82024-520
82024-518EA 379.53 CAD
82024-518 82024-520
VWR® Freezer Racks
Racks Cryogenic Box Racks
Corrosion-resistant, stainless steel horizontal racks feature handles for easy transport.
  • Corrosion-resistant
  • Stainless steel fabrication
  • Multiple shelf counts
Three-shelf rack fits in upright freezers and holds one 7.6 cm (3") box per shelf. Four-shelf rack fits in chest freezers and holds one 5.1 cm (2") box per shelf.
Order Now
SPECIFICATIONS
Environmentally Preferable