VWR® Four-Way Microtube Racks
Supplier: VWR International
82010-750 82010-752 82024-458 82024-460 82024-462 82024-464
82010-750CS 1167.15 CAD
82010-750 82010-752 82024-458 82024-460 82024-462 82024-464
VWR® Four-Way Microtube Racks
Racks Tube Racks
These heavy-duty, polypropylene racks link together to meet custom rack needs.
  • Autoclavable
  • Polypropylene fabrication
Racks feature an interlocking system of tabs and slots for easy connection and sturdy fit. Each rack has four sides that hold either four 50 mL conical tubes, twelve 15 mL conical tubes, thirty-two 1.5 mL microtubes, or thirty-two 0.5 mL microtubes.
Ordering information: Assorted rack pack 82010-750 includes one each in blue, green, pink, yellow, and orange.
Order Now
SPECIFICATIONS
Environmentally Preferable