VWR® SG-900 Semiautomatic Switchover Manifolds for Noncorrosive Gases
Supplier: VWR International
55850-706 55850-708 82023-734 82023-736 82023-738 82023-742 82023-744 82023-746 82023-748 82023-750
55850-706EA 6281.92 CAD
55850-706 55850-708 82023-734 82023-736 82023-738 82023-742 82023-744 82023-746 82023-748 82023-750
VWR® SG-900 Semiautomatic Switchover Manifolds for Noncorrosive Gases
Gas Regulators
The SG-900 system is designed to prevent the depletion of gas during testing and to eliminate the downtime required to change empty cylinders.
  • Automatic switch to reserve gas cylinder
  • Easy to refill cylinders without interrupting work
  • Chrome-plated brass construction
  • Reverse flow check valves
The manifold will automatically switch gas supply from the primary cylinder to a reserve cylinder when the primary supply is depleted
The manifold is chrome-plated brass. All pigtails have a reverse flow check valve to prevent back flow of gas while changingMore Product Information