VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 127 mm Probe
Supplier: VWR International
82021-154
82021-154EA 91.31 CAD
82021-154
VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 127 mm Probe
Thermometers Digital Thermometers
Will read temperatures in liquids, semi-solids and gases.
  • Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialized, CE marked
Certifications: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Includes multi-language instructions (English, French, German,More Product Information