XPert™ Filtered Balance Systems, Labconco®
Supplier: Labconco
3950201 3950202 3950221 3950222 3950301 3950302 3950321 3950322 3950401 3950402 3950421 3950422 3950200 3950220 3950300 3950320 3950400 3950420 3950500 3950501 3950502 3950520 3950521 3950522 3950530 3950531 3950532 3950540 3950541 3950542 3950600 3950601 3950602 3950620 3950621 3950622 3950630 3950631 3950632 3950640 3950641 3950642
82018-108EA 13840.8 CAD
82018-108 82018-110 82018-112 82018-114 82018-116 82018-118 82018-120 82018-122 82018-124 82018-126 82018-128 82018-130 89004-482 89004-484 89004-486 89004-488 89004-490 89004-492 93000-516 93000-518 93000-520 93000-522 93000-524 93000-526 93000-528 93000-530 93000-532 93000-534 93000-536 93000-538 93000-540 93000-542 93000-544 93000-546 93000-548 93000-550 93000-552 93000-554 93000-556 93000-558 93000-560 93000-562
XPert™ Filtered Balance Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
These portable systems provide user protection by keeping powders and particulates contained during weighing procedures
The deep interior easily accommodates large micro and analytical balances. The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. The upper containment sash foil bleeds air into the enclosure to direct concentrations of contaminants away from the operator’s breathing zone. The upper dilution air supply introduces room air at the top of the sash toMore Product Information