Stirring Elements, IKA® Works
Supplier: IKA Works
0000741000 0002333000 0000741300 0001295900 0002333100 0002387900 0000756900 0001132700 0757700 0000739000 0000739500 0002747400 0000477700 0000477800 0001296000 0002343600 0000513500 0002022300
33994-032EA 1558.24 CAD
33994-032 33994-074 33994-112 33994-116 33994-122 33994-126 33994-130 33994-132 33994-136 82017-852 82017-854 82017-856 89176-704 89176-706 33994-114 33994-118 33994-124 33994-138
Stirring Elements, IKA® Works
Stirrers Stirring Tools
Made of stainless steel
Accepted by overhead stirrer motors with corresponding output shaft/chuck diameter measurements. Four-bladed propeller stirrers generate axial flow, drawing media from top to bottom, and local shearing forces. Three-bladed propellers produce efficient flow with minimal shearing. Turbine stirrers generate axial flow, pulling material from above, with minimal shearing. Configuration reduces the risk of user injury if blade contacts the vessel wall.
Dissolver stirrers produce radial flow, drawing material from both top and bottom, with high turbulence and shearingMore Product Information