VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Supplier: VWR International
82021-118 82021-122 82007-142 82007-144 82007-146 82007-148 82007-150 82007-152 82007-154 82007-156 82007-158 82007-160 82007-162 82021-114 82021-116 82021-120 82021-124 82021-126 82021-128 97014-322 89214-736 89214-738 89214-740 89214-742 89214-744
82021-118PK 35.11 CAD
82021-118 82021-122 82007-142 82007-144 82007-146 82007-148 82007-150 82007-152 82007-154 82007-156 82007-158 82007-160 82007-162 82021-114 82021-116 82021-120 82021-124 82021-126 82021-128 97014-322 89214-736 89214-738 89214-740 89214-742 89214-744
VWR® CryoPro® Fiberboard Storage Boxes and Dividers
Boxes Cryoboxes
Boxes are constructed of durable fiberboard with a protective, moisture-repellant coating.
  • Available with or without slots or dividers
Liquid nitrogen (LN2) freezer boxes feature slots for safe LN2 drainage and reduced LN2 consumption. Mechanical freezer boxes do not feature slots and cannot be used with LN2. Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accomodate dividers (sold separately) for 16 to 100 cells. Fiberboard dividers can fit in any 13.2Wx13.2Lcm (51/5x51/5" ) LN2More Product Information