Distillation Grid Kits for Protector®, Premier™ and XStream® Fume Hoods, Labconco®
Supplier: Labconco
9725204 9725205 9725206 9725200
82006-970EA 1651.1 CAD
82006-970 82006-972 82006-974 82006-976
Distillation Grid Kits for Protector®, Premier™ and XStream® Fume Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
These kits let the user mount glassware and other small apparatus to the rear of the hood’s interior, above the work surface to maximize usable working area.
Ordering information: Kits include all of the stainless steel rods and connectors required for installation. Easy-to-understand instruction sheet is also included. Items are mounted to the grid with customer-supplied clamps and supports; contact your Avantor Furniture Specialist for information.
Order Now
SPECIFICATIONS
Environmentally Preferable